Methyl-5-methylsalicylat

Lieferant: CHEMPUR

Synonyme: Methyl 2-hydroxy-5-methylbenzoate

RF01204.250 RF01204.1
CHMPRF01204.250EA 0 EUR
CHMPRF01204.250 MAYBRF01204.1
Methyl-5-methylsalicylat
Methyl-5-methylsalicylat
Formel: C₉H₁₀O₃
Molecular Weight: 166,1700 g/mol
MDL: MFCD00016485
CAS-Nummer: 22717-57-3
EINECS: 245-173-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR