Methyl-4-(hydroxymethyl)benzoat ≥99%

Lieferant: Thermo Scientific

Synonyme: Methyl 4-(hydroxymethyl)benzoate

B22564.06 B22564.14
B22564.06EA 47.3 EUR
B22564.06 B22564.14
Methyl-4-(hydroxymethyl)benzoat ≥99%
Methyl-4-(hydroxymethyl)benzoat

Purity: 99% CAS No.: 6908-41-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 46-52° MDL No.: MFCD00009984

Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 46…52 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009984
CAS-Nummer: 6908-41-4
EINECS: 230-017-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR