2-Fluor-5-(methoxycarbonylmethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester

PN-5703-1G PN-5703-250M
COBBPN-5703-1GEA 132 EUR
COBBPN-5703-1G COBBPN-5703-250M
2-Fluor-5-(methoxycarbonylmethyl)phenylboronsäurepinakolester
2-Fluor-5-(methoxycarbonylmethyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₀BFO₄
Molecular Weight: 294.13 g/mol
MDL: MFCD12026092
CAS-Nummer: 944317-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR