1-Methyl-4-(trifluormethylsulfonyl)benzol

Lieferant: COMBI-BLOCKS

OT-0801-5G OT-0801-1G
COBBOT-0801-5GEA 2510 EUR
COBBOT-0801-5G COBBOT-0801-1G
1-Methyl-4-(trifluormethylsulfonyl)benzol
1-Methyl-4-(trifluormethylsulfonyl)benzol
Formel: C₈H₇F₃O₂S
MDL: MFCD00523749
CAS-Nummer: 383-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR