((Cyclopentyloxy)methyl)benzol

Lieferant: COMBI-BLOCKS

OT-1211-5G OT-1211-1G
COBBOT-1211-5GEA 451 EUR
COBBOT-1211-5G COBBOT-1211-1G
((Cyclopentyloxy)methyl)benzol
((Cyclopentyloxy)methyl)benzene
Formel: C₁₂H₁₆O
MDL: MFCD16295098
CAS-Nummer: 40843-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR