4-Benzyloxy-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Benzyloxy-2-methylbenzeneboronic acid

BB-2682-5G BB-2682-25G BB-2682-1G
COBBBB-2682-5GEA 262 EUR
COBBBB-2682-5G COBBBB-2682-25G COBBBB-2682-1G
4-Benzyloxy-2-methylphenylboronsäure
4-Benzyloxy-2-methylphenylboronsäure
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242.08 g/mol
MDL: MFCD03788404
CAS-Nummer: 847560-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR