4-Benzyloxy-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Benzyloxy)-3-methylphenyl)boronic acid, 4-Benzyloxy-3-methylbenzeneboronic acid

BB-2744-1G BB-2744-5G BB-2744-25G
COBBBB-2744-1GEA 125 EUR
COBBBB-2744-1G COBBBB-2744-5G COBBBB-2744-25G
4-Benzyloxy-3-methylphenylboronsäure
4-Benzyloxy-3-methylphenylboronsäure
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242.08 g/mol
MDL: MFCD06409944
CAS-Nummer: 338454-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR