3-Benzyloxy-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Benzyloxy)-5-methylphenyl)boronic acid, 3-(Benzyloxy)-5-methylphenylboronic acid

FA-2867-1G FA-2867-250M
COBBFA-2867-1GEA 595 EUR
COBBFA-2867-1G COBBFA-2867-250M
3-Benzyloxy-5-methylphenylboronsäure
3-Benzyloxy-5-methylphenylboronsäure
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242,0800 g/mol
MDL: MFCD16295086
CAS-Nummer: 1256355-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR