3-Brom-2-(3'-methoxybenzyloxy)-5-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8221-1G
BB-8221-250M
COBBBB-8221-1GEA
303
EUR
COBBBB-8221-1G
COBBBB-8221-250M
3-Brom-2-(3'-methoxybenzyloxy)-5-methylphenylboronsäure
3-Brom-2-(3'-methoxybenzyloxy)-5-methylphenylboronsäure
Formel:
C₁₅H₁₆BBrO₄ |
MDL:
MFCD06411315 CAS-Nummer: 849062-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...