3-Brom-2-(3-chlorbenzyloxy)-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-2-((3-chlorobenzyl)oxy)-5-methylphenyl)boronic acid

BB-8118-1G BB-8118-250M
COBBBB-8118-1GEA 451 EUR
COBBBB-8118-1G COBBBB-8118-250M
3-Brom-2-(3-chlorbenzyloxy)-5-methylphenylboronsäure
3-Brom-2-(3-chlorbenzyloxy)-5-methylphenylboronsäure
Formel: C₁₄H₁₃BBrClO₃
Molecular Weight: 355.42 g/mol
MDL: MFCD06798105
CAS-Nummer: 870778-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR