3-Brom-2-(4-chlorbenzyloxy)-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-2-((4-chlorobenzyl)oxy)-5-methylphenyl)boronic acid

BB-8055-1G BB-8055-250M
COBBBB-8055-1GEA 470 EUR
COBBBB-8055-1G COBBBB-8055-250M
3-Brom-2-(4-chlorbenzyloxy)-5-methylphenylboronsäure
3-Brom-2-(4-chlorbenzyloxy)-5-methylphenylboronsäure
Formel: C₁₄H₁₃BBrClO₃
Molecular Weight: 355,4200 g/mol
MDL: MFCD06411365
CAS-Nummer: 849052-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR