Nordihydroguajaretsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L03149.06 L03149.03
L03149.06EA 268 EUR
L03149.06 L03149.03
Nordihydroguajaretsäure ≥97%
Nordihydroguajaretsäure

Purity: 97% CAS No.: 500-38-9 Molecular Formula: C18H22O4 Formula Weight: 302.37 Odour: Odourless Melting Point: 184-188° Beilstein Reference: 2056825 Merck Reference: 14,6693 MDL No.: MFCD00002206

Formel: C₁₈H₂₂O₄
Molecular Weight: 302.37 g/mol
Schmelzpunkt: 184…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002206
CAS-Nummer: 500-38-9
EINECS: 207-903-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR