Nordihydroguajaretsäure, Sigma-Aldrich®
Lieferant: Merck
74540-1G
74540-5G
SIAL74540-1GEA
89.3
EUR
SIAL74540-1G
SIAL74540-5G
Nordihydroguajaretsäure, Sigma-Aldrich®
Nordihydroguajaretsäure
Formel:
C₁₈H₂₂O₄ Molecular Weight: 302,37 g/mol |
MDL:
MFCD00002206 CAS-Nummer: 500-38-9 EINECS: 207-903-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...