(±)-Cbz-α-phosphonoglycintrimethylester ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate
B22506.06
B22506.14
B22506.06EA
79.3
EUR
B22506.06
B22506.14
(±)-Cbz-α-phosphonoglycintrimethylester ≥97%
(±)-Cbz-α-phosphonoglycintrimethylester
Purity: 97%
CAS No.: 88568-95-0
Molecular Formula: C13H18NO7P
Formula Weight: 331.27
Melting Point: 76-80°
MDL No.: MFCD00043304
Formel:
C₁₃H₁₈NO₇P Molecular Weight: 331,27 g/mol Schmelzpunkt: 76…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043304 CAS-Nummer: 88568-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...