1-Brom-2-(cyclopropylmethoxy)benzol

Lieferant: COMBI-BLOCKS

OT-1121-5G OT-1121-1G
COBBOT-1121-5GEA 1110 EUR
COBBOT-1121-5G COBBOT-1121-1G
1-Brom-2-(cyclopropylmethoxy)benzol
1-Bromo-2-(cyclopropylmethoxy)benzene
Formel: C₁₀H₁₁BrO
MDL: MFCD12138831
CAS-Nummer: 494773-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR