1-Brom-3-(cyclopropylmethoxy)benzol

Lieferant: COMBI-BLOCKS

OT-1156-5G OT-1156-1G
COBBOT-1156-5GEA 884 EUR
COBBOT-1156-5G COBBOT-1156-1G
1-Brom-3-(cyclopropylmethoxy)benzol
1-Bromo-3-(cyclopropylmethoxy)benzene
Formel: C₁₀H₁₁BrO
Molecular Weight: 227,1000 g/mol
MDL: MFCD12138830
CAS-Nummer: 126909-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR