3-(Benzylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: [3-(Benzylamino-1-carbonyl)phenyl]boronic acid pinacol ester

PN-3055-5G PN-3055-1G
COBBPN-3055-5GEA 0 EUR
COBBPN-3055-5G COBBPN-3055-1G
3-(Benzylcarbamoyl)phenylboronsäurepinakolester
3-(Benzylcarbamoyl)phenylboronsäurepinakolester
Formel: C₂₀H₂₄BNO₃
MDL: MFCD09266183
CAS-Nummer: 1048647-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR