4,4'-Biphenyldiboronsäurebis(pinakol)ester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

PN-8747-5G PN-8747-1G
COBBPN-8747-5GEA 249 EUR
COBBPN-8747-5G COBBPN-8747-1G
4,4'-Biphenyldiboronsäurebis(pinakol)ester
4,4'-Biphenyldiboronsäurebis(pinakol)ester
Formel: C₂₄H₃₂B₂O₄
Molecular Weight: 406,14 g/mol
MDL: MFCD08669545
CAS-Nummer: 207611-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR