3-Brom-2-(3'-methoxybenzyloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Bromo-2-((3-methoxybenzyl)oxy)phenyl)boronic acid
BB-8198-1G
BB-8198-250M
COBBBB-8198-1GEA
451
EUR
COBBBB-8198-1G
COBBBB-8198-250M
3-Brom-2-(3'-methoxybenzyloxy)phenylboronsäure
3-Brom-2-(3'-methoxybenzyloxy)phenylboronsäure
Formel:
C₁₄H₁₄BBrO₄ Molecular Weight: 336.970 g/mol |
MDL:
MFCD06411374 CAS-Nummer: 849052-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...