3-Brom-2-(4'-chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8146-1G BB-8146-250M
COBBBB-8146-1GEA 375 EUR
COBBBB-8146-1G COBBBB-8146-250M
3-Brom-2-(4'-chlorbenzyloxy)phenylboronsäure
3-Brom-2-(4'-chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BBrClO₃
MDL: MFCD06411373
CAS-Nummer: 849052-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR