3-Chlor-4-(3'-brombenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8115-1G BB-8115-250M
COBBBB-8115-1GEA 215 EUR
COBBBB-8115-1G COBBBB-8115-250M
3-Chlor-4-(3'-brombenzyloxy)phenylboronsäure
3-Chlor-4-(3'-brombenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BBrClO₃
MDL: MFCD06411317
CAS-Nummer: 849062-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR