2-Benzyloxy-5-brom-1,3-dichlorbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dichloro-4-benzyloxybromobenzene

OT-0815-5G OT-0815-1G
COBBOT-0815-5GEA 451 EUR
COBBOT-0815-5G COBBOT-0815-1G
2-Benzyloxy-5-brom-1,3-dichlorbenzol
2-Benzyloxy-5-brom-1,3-dichlorbenzol
Formel: C₁₃H₉BrCl₂O
MDL: MFCD11617257
CAS-Nummer: 155891-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR