1-(Benzyloxy)-4-brom-2,3-difluorbenzol

Lieferant: COMBI-BLOCKS

OT-0505-5G OT-0505-1G
COBBOT-0505-5GEA 0 EUR
COBBOT-0505-5G COBBOT-0505-1G
1-(Benzyloxy)-4-brom-2,3-difluorbenzol
1-(Benzyloxy)-4-brom-2,3-difluorbenzol
Formel: C₁₃H₉BrF₂O
MDL: MFCD09475823
CAS-Nummer: 941294-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR