4-Benzyloxy-1-brom-2-fluorbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Benzyloxy)-1-bromo-2-fluorobenzene

OT-0582-25G OT-0582-5G OT-0582-1G
COBBOT-0582-25GEA 262 EUR
COBBOT-0582-25G COBBOT-0582-5G COBBOT-0582-1G
4-Benzyloxy-1-brom-2-fluorbenzol
4-Benzyloxy-1-brom-2-fluorbenzol
Formel: C₁₃H₁₀BrFO
Molecular Weight: 281,13 g/mol
MDL: MFCD09801041
CAS-Nummer: 185346-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR