4-Brom-3-phenylanilin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromo-[1,1'-biphenyl]-3-amine

AN-1387-1G AN-1387-5G
COBBAN-1387-1GEA 303 EUR
COBBAN-1387-1G COBBAN-1387-5G
4-Brom-3-phenylanilin
4-Brom-3-phenylanilin
Formel: C₁₂H₁₀BrN
Molecular Weight: 248,1185 g/mol
MDL: MFCD11504836
CAS-Nummer: 1036750-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR