Methyl 4-(4-formylphenyl)benzoate, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4'-Formylbiphenyl-4-carbonsäuremethylester
M-Clarity™ Qualitätsstufe = MQ100
650099-1G
650099-5G
SIAL650099-1GEA
90.8
EUR
SIAL650099-1G
SIAL650099-5G
Methyl 4-(4-formylphenyl)benzoate, Sigma-Aldrich®
Methyl 4-(4-formylphenyl)benzoate
Formel:
C₁₅H₁₂O₃ Molecular Weight: 240,25 g/mol |
MDL:
MFCD04039123 CAS-Nummer: 70916-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...