2'-Formylbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4858-5G
CA-4858-1G
COBBCA-4858-5GEA
0
EUR
COBBCA-4858-5G
COBBCA-4858-1G
2'-Formylbiphenyl-3-carbonsäure
2'-Formylbiphenyl-3-carbonsäure
Formel:
C₁₄H₁₀O₃ |
MDL:
MFCD03424616 CAS-Nummer: 205871-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...