2'-Formylbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-Formyl-[1,1'-biphenyl]-4-carboxylic acid

014269-1G 014269-500MG
FLUO014269-1GEA 249 EUR
FLUO014269-1G FLUO014269-500MG
2'-Formylbiphenyl-4-carbonsäure
2'-Formylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₀O₃
Molecular Weight: 226,2300 g/mol
MDL: MFCD03424615
CAS-Nummer: 205871-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR