3-Chlor-4-(3',5'-dimethoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-((3,5-dimethoxybenzyl)oxy)phenyl)boronic acid

BB-8222-1G BB-8222-250M
COBBBB-8222-1GEA 243 EUR
COBBBB-8222-1G COBBBB-8222-250M
3-Chlor-4-(3',5'-dimethoxybenzyloxy)phenylboronsäure
3-Chlor-4-(3',5'-dimethoxybenzyloxy)phenylboronsäure
Formel: C₁₅H₁₆BClO₅
Molecular Weight: 322.550 g/mol
MDL: MFCD06411316
CAS-Nummer: 849062-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR