2-Benzyloxy-1-chlor-3-fluorbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Benzyloxy)-1-chloro-3-fluorobenzene

OT-0993-1G OT-0993-5G
COBBOT-0993-1GEA 98.3 EUR
COBBOT-0993-1G COBBOT-0993-5G
2-Benzyloxy-1-chlor-3-fluorbenzol
2-Benzyloxy-1-chlor-3-fluorbenzol
Formel: C₁₃H₁₀ClFO
MDL: MFCD13195733
CAS-Nummer: 938180-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR