4-(4-Chlorphenyl)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 4'-Chloro-[1,1'-biphenyl]-4-amine, 4-Amino-4'-chlorobiphenyl

OT-8005-5G OT-8005-1G
COBBOT-8005-5GEA 1170 EUR
COBBOT-8005-5G COBBOT-8005-1G
4-(4-Chlorphenyl)anilin
4-(4-Chlorphenyl)anilin
Formel: C₁₂H₁₀ClN
Molecular Weight: 203.6700 g/mol
MDL: MFCD00477989
CAS-Nummer: 135-68-2
EINECS: 205-211-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR