3'-Acetylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4870-1G CA-4870-250M
COBBCA-4870-1GEA 510 EUR
COBBCA-4870-1G COBBCA-4870-250M
3'-Acetylbiphenyl-3-carbonsäure
3'-Acetylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₂O₃
MDL: MFCD03839958
CAS-Nummer: 728918-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR