4'-Acetylbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4841-5G
CA-4841-1G
COBBCA-4841-5GEA
1180
EUR
COBBCA-4841-5G
COBBCA-4841-1G
4'-Acetylbiphenyl-3-carbonsäure
4'-Acetylbiphenyl-3-carbonsäure
Formel:
C₁₅H₁₂O₃ |
MDL:
MFCD04039045 CAS-Nummer: 635757-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...