2-Fluor-4-biphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2723-5G PN-2723-1G
COBBPN-2723-5GEA 595 EUR
COBBPN-2723-5G COBBPN-2723-1G
2-Fluor-4-biphenylboronsäurepinakolester
2-Fluor-4-biphenylboronsäurepinakolester
Formel: C₁₈H₂₀BFO₂
Molecular Weight: 298.160 g/mol
MDL: MFCD09258651
CAS-Nummer: 269410-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR