3-Benzyloxy-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Benzyloxy)-5-(trifluoromethoxy)phenyl)boronic acid

BB-6360-1G BB-6360-5G
COBBBB-6360-1GEA 303 EUR
COBBBB-6360-1G COBBBB-6360-5G
3-Benzyloxy-5-(trifluormethoxy)phenylboronsäure
3-Benzyloxy-5-(trifluormethoxy)phenylboronsäure
Formel: C₁₄H₁₂BF₃O₄
MDL: MFCD16036111
CAS-Nummer: 1256345-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR