2,6-Difluor-3-(3',5'-dimethoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((3,5-Dimethoxybenzyl)oxy)-2,6-difluorophenyl)boronic acid

BB-8179-1G BB-8179-250M
COBBBB-8179-1GEA 341 EUR
COBBBB-8179-1G COBBBB-8179-250M
2,6-Difluor-3-(3',5'-dimethoxybenzyloxy)phenylboronsäure
2,6-Difluor-3-(3',5'-dimethoxybenzyloxy)phenylboronsäure
Formel: C₁₅H₁₅BF₂O₅
Molecular Weight: 324.08 g/mol
MDL: MFCD07369738
CAS-Nummer: 849062-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR