3-Phenylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Biphenylamine hydrochloride, 3-Aminobiphenyl hydrochloride, [1,1'-Biphenyl]-3-amine hydrochloride

AN-1528-5G AN-1528-1G
COBBAN-1528-5GEA 89.5 EUR
COBBAN-1528-5G COBBAN-1528-1G
3-Phenylanilin Hydrochlorid
3-Phenylanilin Hydrochlorid
Formel: C₁₂H₁₂ClN
Molecular Weight: 205,69 g/mol
MDL: MFCD00068358
CAS-Nummer: 2113-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR