Biphenyl-4-sulfonylchlorid

Lieferant: abcr

Synonyme: Biphenyl-4-sulphonyl Chloride

21031.0005 21031.0025
ABCR21031.0005EA 0 EUR
ABCR21031.0005 ABCR21031.0025
Biphenyl-4-sulfonylchlorid
Biphenyl-4-sulfonylchlorid
Formel: C₁₂H₉ClO₂S
Molecular Weight: 252.72 g/mol
MDL: MFCD00078658
CAS-Nummer: 1623-93-4
EINECS: 216-609-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR