8-Fluorchinolin
Lieferant: COMBI-BLOCKS
PY-7377-1G
PY-7377-5G
PY-7377-25G
COBBPY-7377-1GEA
38.5
EUR
COBBPY-7377-1G
COBBPY-7377-5G
COBBPY-7377-25G
8-Fluorchinolin
8-Fluoroquinoline
Formel:
C₉H₆FN Molecular Weight: 147.1500 g/mol |
MDL:
MFCD01685514 CAS-Nummer: 394-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...