5-Amino-8-(trifluormethoxy)chinolin

Lieferant: COMBI-BLOCKS

HC-2111-5G HC-2111-1G
COBBHC-2111-5GEA 0 EUR
COBBHC-2111-5G COBBHC-2111-1G
5-Amino-8-(trifluormethoxy)chinolin
5-Amino-8-(trifluoromethoxy)quinoline
Formel: C₁₀H₇F₃N₂O
MDL: MFCD11504893
CAS-Nummer: 1072944-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR