6-Fluor-2-methoxychinolin

Lieferant: COMBI-BLOCKS

PY-1380-1G PY-1380-5G
COBBPY-1380-1GEA 149 EUR
COBBPY-1380-1G COBBPY-1380-5G
6-Fluor-2-methoxychinolin
6-Fluoro-2-methoxyquinoline
Formel: C₁₀H₈FNO
MDL: MFCD16295129
CAS-Nummer: 1226808-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR