6-Fluor-8-methoxychinolin

Lieferant: COMBI-BLOCKS

HC-3036-1G HC-3036-5G
COBBHC-3036-1GEA 303 EUR
COBBHC-3036-1G COBBHC-3036-5G
6-Fluor-8-methoxychinolin
6-Fluoro-8-methoxyquinoline
Formel: C₁₀H₈FNO
MDL: MFCD15143547
CAS-Nummer: 887769-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR