5,7-Dichlor-2-(trifluormethyl)-4-chinolinol

Lieferant: FLUOROCHEM

Synonyme: 5,7-Dichloro-4-hydroxy-2-(trifluoromethyl)quinoline

004720-1G 004720-5G
FLUO004720-1GEA 72.9 EUR
FLUO004720-1G FLUO004720-5G
5,7-Dichlor-2-(trifluormethyl)-4-chinolinol
5,7-Dichlor-2-(trifluormethyl)-4-chinolinol
Formel: C₁₀H₄Cl₂F₃NO
Molecular Weight: 282.0500 g/mol
MDL: MFCD00173851
CAS-Nummer: 59108-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR