6,8-Dichlor-2-(trifluormethyl)-4-chinolinol

Lieferant: COMBI-BLOCKS

HC-6752-5G HC-6752-1G
COBBHC-6752-5GEA 0 EUR
COBBHC-6752-5G COBBHC-6752-1G
6,8-Dichlor-2-(trifluormethyl)-4-chinolinol
6,8-Dichlor-2-(trifluormethyl)-4-chinolinol
Formel: C₁₀H₄Cl₂F₃NO
MDL: MFCD00601299
CAS-Nummer: 18706-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR