2-Methyl-2-propanyl-(trans-4-hydroxycyclohexyl)carbamat

Lieferant: abcr

Synonyme: tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected

TYB50390 TYB50390.0005
ABCRTYB50390EA 0 EUR
ABCRTYB50390 ABCRTYB50390.0005
2-Methyl-2-propanyl-(trans-4-hydroxycyclohexyl)carbamat
trans-4-(Boc-amino)cyclohexanol
Formel: C₁₁H₂₁NO₃
Molecular Weight: 215.28 g/mol
MDL: MFCD03844613
CAS-Nummer: 111300-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR