trans-4-(Dimethylamino)cyclohexanol

Lieferant: COMBI-BLOCKS

AM-1615-5G AM-1615-1G
COBBAM-1615-5GEA 1220 EUR
COBBAM-1615-5G COBBAM-1615-1G
trans-4-(Dimethylamino)cyclohexanol
trans-4-(Dimethylamino)cyclohexanol
Formel: C₈H₁₇NO
MDL: MFCD09258736
CAS-Nummer: 103023-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR