1-(Nitromethyl)cyclohexen

Lieferant: Thermo Scientific

JFD00082.250 JFD00082.1
MAYBJFD00082.250EA 0 EUR
MAYBJFD00082.250 MAYBJFD00082.1
1-(Nitromethyl)cyclohexen
1-(Nitromethyl)cyclohexene
Formel: C₇H₁₁NO₂
CAS-Nummer: 5330-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR