Kalium-3-(trifluoroborato)propionatethylester

Lieferant: COMBI-BLOCKS

Synonyme: Potassium (3-ethoxy-3-oxopropyl)trifluoroborate

TB-0014-1G TB-0014-5G
COBBTB-0014-1GEA 63.4 EUR
COBBTB-0014-1G COBBTB-0014-5G
Kalium-3-(trifluoroborato)propionatethylester
Kalium-3-(trifluoroborato)propionatethylester
Formel: C₅H₉BF₃KO₂
Molecular Weight: 208,300 g/mol
MDL: MFCD09993030
CAS-Nummer: 1023357-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR