Kalium-1,3-dimethyluracil-5-trifluoroborat

Lieferant: COMBI-BLOCKS

TB-0008-1G TB-0008-5G
COBBTB-0008-1GEA 420 EUR
COBBTB-0008-1G COBBTB-0008-5G
Kalium-1,3-dimethyluracil-5-trifluoroborat
Kalium-1,3-dimethyluracil-5-trifluoroborat
Formel: C₆H₇BF₃KN₂O₂
MDL: MFCD09271782
CAS-Nummer: 1150654-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR