Ethylphenoxyacetat-4-trifluoroborat Kaliumsalz
Lieferant: COMBI-BLOCKS
Synonyme:
Potassium (4-(2-ethoxy-2-oxoethoxy)phenyl)trifluoroborate
TB-0003-1G
TB-0003-5G
COBBTB-0003-1GEA
149
EUR
COBBTB-0003-1G
COBBTB-0003-5G
Ethylphenoxyacetat-4-trifluoroborat Kaliumsalz
Ethylphenoxyacetat-4-trifluoroborat Kaliumsalz
Formel:
C₁₀H₁₁BF₃KO₃ Molecular Weight: 286,1 g/mol |
MDL:
MFCD09260451 CAS-Nummer: 1150654-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...